Kronecker Ligature Needle, OR Grade, Sklar®
Supplier: Sklar
26-1610 26-1611
76193-002EA 247.29 USD
76193-002 76193-004
Kronecker Ligature Needle, OR Grade, Sklar®
Dissection Needles
The Sklar® Kronecker Ligature Needle is used for guiding suture material into difficult to reach areas or deep muscle and tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sharp enough for tough tissue while not damaging the surrounding area. The Kronecker is availale with a right or left curve.