ProDeliverIN CRISPR, OZ Biosciences
Supplier: OZ Biosciences
ProDeliverIN CRISPR kit is a transfection reagent optimized for recombinant Cas9 protein delivery or Cas9/gRNA RNP complexes. For your gene editing applications, this reagent provides high transfection efficiency with minimal toxicity.
- High efficiency delivery of Cas9 protein or Cas9 gRNA complexes
- Low cell toxicity - enable to start your experiment with less cells
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). As for mRNA, it present also the advantages of avoiding genomic integration and it’s cell cycle independent.
Efficient delivery represents a critical step for genome editing experiments. For the most efficient Cas9 nuclease delivery using ProDeliverIN CRSIPR, we recommend our Optimized Cas9 nuclease.
Información de suministro: Supplied with 100 µL of R-Phycoerythrin Positive Control.