Dicarbonyl(2,4-pentanedionato)iridium(I) 98%

Supplier: STREM CHEMICALS INC MS
Warning

77-1050-2G 77-1050-500MG
100202-690EA 825.17 USD
100202-690 100206-804
Dicarbonyl(2,4-pentanedionato)iridium(I) 98%
Dicarbonyl(2,4-pentanedionato)iridium(I)
Formula: C₇H₇IrO₄
MDL Number: MFCD00049135
CAS Number: 14023-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR