Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate 98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Cobaltocene Hexafluorophosphate, Cobalticinium hexafluorophosphate

27-0410-1G 27-0410-5G
100196-184EA 108.98 USD
100196-184 100207-378
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate 98%
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₀CoF₆P
MW: 334.09 g/mol
MDL Number: MFCD00011874
CAS Number: 12427-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR