Dog Recombinant RC GM-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
400-14-5UG 400-14-20UG 400-14-100UG 400-14-500UG 400-14-1MG
76199-588EA 82.09 USD
76199-588 76199-590 76199-592 76199-594 76199-596
Dog Recombinant RC GM-CSF (from E. coli)
Proteins and Peptides
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.