GDC-0449 free base ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-Chloro-N-(4-chloro-3-(pyridin-2-yl)phenyl)-4-(methylsulphonyl)benzamide, Vismodegib

CHM242-0500 CHM242-1000 CHM242-2000 CHM242-5000 CHM242-0100 CHM242-0200
76203-376EA 571.16 USD
76203-376 76203-378 76203-380 76203-382 76203-458 76203-460 76203-462
GDC-0449 free base ≥99% (by HPLC)
GDC-0449

Vismodegib is a potent, novel and specific inhibitor of hedgehog with IC50 of 3 nM and also inhibitor of P-gp with IC50 of 3.0 μM.


  • Hedgehog inhibitor
  • Soluble in DMSO at 200 mg/mL

Formula: C₁₉H₁₄Cl₂N₂O₃S
MW: 421.3 g/mol
MDL Number: MFCD12407408
CAS Number: 879085-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=887f0c1fb215178772cf0b2903d16a7b0cb24a4b&vid=ef0225880239eb8f2f84e8732031165fefcf527f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR