GSK1120212 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

CHM239-0010 CHM239-0050 CHM239-0100 CHM239-0200 CHM239-0500 CHM239-1000 CHM239-2000
76203-406EA 92.65 USD
76203-406 76203-408 76203-410 76203-412 76203-414 76203-416 76203-418 76203-420
GSK1120212 ≥99% (by HPLC)
GSK1120212

Trametinib is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays.


  • Potent MEK1/2 inhibitor
  • White solid
  • Soluble in DMSO (20 mg/mL).

Formula: C₂₆H₂₃FIN₅O₄
MW: 615.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17215075
CAS Number: 871700-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57afe0bd6f724d14acde3582529f964970e44849&vid=a1aa2a7d370c1e70d6297881553b68678005ff0b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR