Sonidegib diphosphate ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: rel-N-(6-((2R,6S)-2,6-dimethylmorpholino)pyridin-3-yl)-2-methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-carboxamide bis(phosphate)

CHM237-0010 CHM237-0025 CHM237-0050 CHM237-0100 CHM237-0200 CHM237-0500
76203-280EA 198.38 USD
76203-280 76203-282 76203-284 76203-286 76203-288 76203-290 76203-374
Sonidegib diphosphate ≥99% (by HPLC)
Sonidegib diphosphate

Sonidegib, also known as LDE-225, is a potent and selective smoothened (Smo) antagonist.


  • Smoothened antagonist


White solid.


Soluble in DMSO.

Formula: C₂₆H₃₂F₃N₃O₁₁P₂
MW: 681.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16627994
CAS Number: 1218778-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8ce238bd17f7381452c1ae44e83be81e01a859&vid=021e45f215cac92445720815923251a32d8eccee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR