Rucaparib phosphate ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one phosphate
CHM236-0005
CHM236-0010
CHM236-0025
CHM236-0050
CHM236-0100
CHM236-0200
CHM236-0300
76203-360EA
180.94
USD
76203-360
76203-362
76203-364
76203-366
76203-368
76203-370
76203-372
Rucaparib phosphate ≥99% (by HPLC)
Rucaparib phosphate
Rucaparib is a poly (ADP ribose) polymerase (PARP) inhibitor. PARP is a DNA damage-activated nuclear enzyme that has a key signaling role in the base excision repair pathway.
- PARP inhibitor
Yellow solid.
Soluble in DMSO.
Formula:
C₁₉H₂₁FN₃O₅P MW: 421.3593 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17010269 CAS Number: 459868-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...