Rucaparib free base ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one

CHM235-0005 CHM235-0010 CHM235-0025 CHM235-0050 CHM235-0100 CHM235-0200 CHM235-0300
76203-346EA 182.03 USD
76203-346 76203-348 76203-350 76203-352 76203-354 76203-356 76203-358
Rucaparib free base ≥99% (by HPLC)
Rucaparib

Rucaparib is a poly (ADP ribose) polymerase (PARP) inhibitor. PARP is a DNA damage-activated nuclear enzyme that has a key signaling role in the base excision repair pathway.


  • PARP inhibitor


Yellow solid.


Soluble in DMSO.

Formula: C₁₉H₁₈FN₃O
MW: 323.3641 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11977252
CAS Number: 283173-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=980ea5e1cbd3f348da98daaadd4d06041af807ab&vid=50e99dfd3bc25d2285e5c7c8f1624a442b7a6918&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR