Nintedanib ethanesulfonate salt ≥99% (by HPLC)
Supplier: Enzo Life Sciences
CHM229-0050
CHM229-0100
CHM229-0200
CHM229-0500
CHM229-1000
76203-266EA
136.25
USD
76203-266
76203-268
76203-270
76203-272
76203-274
Nintedanib ethanesulfonate salt ≥99% (by HPLC)
Nintedanib ethanesulfonate salt
Nintedanib is a potent inhibitor of multiple receptor tyrosine kinases (RTKs) and non-receptor tyrosine kinases (nRTKs).
- Potent tyrosine kinase inhibitor
Yellow solid.
Soluble in DMSO (25 mg/mL).
Formula:
C₃₃H₃₉N₅O₇S Storage Temperature: Freezer |
MDL Number:
MFCD26142360 CAS Number: 656247-18-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...