Neratinib ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: HKI-272, (E)-N-(4-((3-Chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide

CHM228-0010 CHM228-0025 CHM228-0050 CHM228-0100 CHM228-0200 CHM228-0500 CHM228-1000
76203-250EA 174.4 USD
76203-250 76203-252 76203-254 76203-256 76203-258 76203-260 76203-262
Neratinib ≥99% (by HPLC)
Neratinib

Neratinib is a potent and selective inhibitor of EGFR and HER2.


  • Potent and selective inhibitor of EGFR and HER2
  • Off-white solid
  • Soluble in DMSO at 2 mg/mL.

Formula: C₃₀H₂₉ClN₆O₃
Storage Temperature: Freezer
MDL Number: MFCD09752958
CAS Number: 698387-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e28f189e692f03e3bed9165c23552a9b2e3ae4&vid=c0c7a3735b50c988a7fba35789bf928a51b47a4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR