Dabrafenib free base ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide

CHM219-0025 CHM219-0050 CHM219-0100 CHM219-0200 CHM219-0500 CHM219-1000 CHM219-2000
76203-144EA 117 USD
76203-144 76203-146 76203-148 76203-150 76203-152 76203-154 76203-156 76203-158
Dabrafenib free base ≥99% (by HPLC)
Dabrafenib

Dabrafenib is a mutant BRAFV600 specific inhibitor with IC50 of 0.8 nM in cell-free assays, with 4- and 6-fold less potency against B-Raf(wt) and c-Raf, respectively.


  • Specific inhibitor of mutant BRAFV600


White solid.


Soluble in DMSO.

Formula: C₂₃H₂₀F₃N₅O₂S₂
Storage Temperature: Freezer
MDL Number: MFCD17215684
CAS Number: 1195765-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a17fc2cc63eccd403cd4383b1421567bce2306&vid=523e918ec181939e9ef20d3e34c196130c56f06a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR