LDK378 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: LDK-378, Ceritinib (free base), 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulphonyl)phenyl)pyrimidine-2,4-diamine

CHM218-0005 CHM218-0010 CHM218-0025 CHM218-0050 CHM218-0100 CHM218-0200 CHM218-0500
76203-128EA 173.31 USD
76203-128 76203-130 76203-132 76203-134 76203-136 76203-138 76203-140
LDK378 ≥99% (by HPLC)
LDK378

Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer.


  • Selective ALK inhibitor
  • Off-white solid
  • Soluble in DMSO.

Formula: C₂₈H₃₆ClN₅O₃S
MW: 558.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26142648
CAS Number: 1032900-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=400c512ebf59a2a9dd6520f451ccac811db20ea7&vid=455f3f4d06cdfabc83fee178ac5215af132aa822&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR