Alectinib hydrochloride ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
9-Ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[B]carbazole-3-carbonitrile,hydrochloride
CHM213-0025
CHM213-0050
CHM213-0100
CHM213-0200
CHM213-0250
CHM213-0500
76203-494EA
406.57
USD
76203-494
76203-496
76203-498
76203-500
76203-502
76203-504
Alectinib hydrochloride ≥99% (by HPLC)
Alectinib hydrochloride
Alectinib hydrochloride is a selective anaplastic lymphoma kinase (ALK) inhibitor leading to inhibition of tumor cell proliferation and induction of cell death.
- ALK inhibitor
Soluble in DMSO.
Formula:
C₃₀H₃₅ClN₄O₂ MW: 519.08 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD27987893 CAS Number: 1256589-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...