Renilla Luciferase Assay System, Promega
Supplier: Promega Corporation
E2810 E2820
PAE2810PK 211.23 USD
PAE2810 PAE2820
Renilla Luciferase Assay System, Promega
Assays Enzyme Assays
Renilla Luciferase Assay System provides a fast, sensitive method for detecting luciferase from sea pansy (Renilla reniformis).
  • Fast, Sensitive Detection of Renilla Luciferase Activity
  • Linear range extends over 7 logs
  • Low background, increased sensitivity
  • Detect 10-19 moles of Renilla luciferase
Renilla Luciferase Assay System is designed to provide a fast and sensitive method of detecting luciferase from sea pansy (Renilla reniformis). The system is a convenient alternative to firefly (Photinus pyralis) reporter systems and is designed to yield reliable, linear results for a concentration range over 7 orders of magnitude. The Renilla Luciferase Assay System has been formulated with a proprietary composition that significantly reduces the effect of coelenterazine autoluminescence when compared to other reagents, making the reagent orders of magnitude more sensitive than published methods. This system enables measurements with wildtype and synthetic hRluc genes for primary expression or internal normalization measurements of gene expression.