NanoBRET Nano-Glo Substrate, Promega
Supplier: Promega Corporation
N1571 N1572 N1573
PAN1571EA 475.91 USD
PAN1571 PAN1572 PAN1573
NanoBRET Nano-Glo Substrate, Promega
Assays Cellular Assays
The NanoBRET™ Nano-Glo(R) Substrate is the substrate used by NanoLuc(R) Luciferase to generate the donor signal in several NanoBRET™ Assays.