2-Chloro-6-(trichloromethyl)pyridine ≥99%
Supplier: CHEM IMPEX INTL INC MS
|
Danger
|
Synonyms:
Nitrapyrin
27826-5G
27826-25G
27826-1G
102560-990EA
105.09
USD
102560-990
102585-440
102604-972
2-Chloro-6-(trichloromethyl)pyridine ≥99%
2-Chloro-6-(trichloromethyl)pyridine
Formula:
C₆H₃Cl₄N MW: 230.897 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00072500 CAS Number: 1929-82-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity | ≥99% |
Water Content (Karl Fisher) | ≤0.1% |
TCMP | ≤0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...