Fmoc-3-(trifluoromethyl)-L-phenylalanine ≥99% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Fmoc-Phe(3-CF₃)-OH, Fmoc-Phe(3-CF3)-OH, (S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-3-(3-trifluoromethylphenyl)propionic acid, Fmoc-L-Phe-(3-trifluoromethyl)-OH, Fmoc-L-Phe-(3-CF3)-OH, 3-(Trifluoromethyl)-L-phenylalanine, N-FMOC protected

07386-1G 07386-5G 07386-25G
102566-692EA 169.11 USD
102566-692 102607-496 102630-096
Fmoc-3-(trifluoromethyl)-L-phenylalanine ≥99% (by HPLC)
Fmoc-3-(trifluoromethyl)-L-phenylalanine

Soluble in DMF (0.3 g in 2 ml).

Formula: C₂₅H₂₀F₃NO₄
MW: 455.44 g/mol
Melting Pt: 126...133 °C
Storage Temperature: Freezer
MDL Number: MFCD00672555
CAS Number: 205526-27-8

Order Now

Specification Test Results

Optical Rotation, [a]D20 –35 ±2° (C=1 in DMF)
Infrared Spectrum Conforms to Structure
Analysis by Mass Spectra Conforms to Standard
Loss on Drying ≤2.0% (70 °C , 2 h)
¹H NMR Conforms to Structure
Purity (by HPLC) ≥99%
Water Content (Karl Fisher) ≤1%
Chiral HPLC ≥99.5% (ee)
D-Enantiomer ≤0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR