N-Boc-L-alanine ≥98% (by HPLC), white to off-white powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

01322-25G 01322-1KG 01322-100G 01322-250G
102573-460EA 28.83 USD
102573-460 102588-200 102615-970 102615-972
N-Boc-L-alanine ≥98% (by HPLC), white to off-white powder
N-Boc-L-alanine
Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Melting Pt: 76…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037225
CAS Number: 15761-38-3

Order Now

Specification Test Results

Optical Rotation, [a]D –25 ±2° (C=1 in AcOH)
Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR