Bis(dibenzylideneacetone)palladium, purple powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bis(dibenzylideneacetone)palladium(0)
46-0210-1G
46-0210-250MG
46-0210-5G
46-0210-25G
100203-186EA
80.68
USD
100203-186
100203-184
100203-188
102978-552
Bis(dibenzylideneacetone)palladium, purple powder
Bis(dibenzylideneacetone)palladium
- Purple color
- Air and moisture sensitive
Formula:
C₃₄H₂₈O₂Pd MW: 575 g/mol |
MDL Number:
MFCD00051942 CAS Number: 32005-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...