Sodium selenite pentahydrate ≥99.0% (RT), Puriss. p.a., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
00163-100G
00163-25G
BJ00163-100GEA
249
USD
BJ00163-100G
BJ00163-25G
Sodium selenite pentahydrate ≥99.0% (RT), Puriss. p.a., Fluka™
Sodium selenite pentahydrate
Formula:
Na₂O₃Se·5H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00149173 CAS Number: 26970-82-1 EINECS: 233-267-9 UN: 2630 ADR: 6.1,I |
Specification Test Results
Assay | 99.0 - 101.0 % |
Calcium (Ca) | max. 20 ppm |
Cadmium (Cd) | max. 5 ppm |
Cobalt (Co) | max. 5 ppm |
Chromium (Cr) | max. 5 ppm |
Copper (Cu) | max. 5 ppm |
Iron (Fe) | max. 10 ppm |
Potassium (K) | max. 50 ppm |
Magnesium (Mg) | max. 5 ppm |
Manganese (Mn) | max. 5 ppm |
Nickel (Ni) | max. 5 ppm |
Lead (Pb) | max. 5 ppm |
Zinc (Zn) | max. 5 ppm |
Chloride (Cl) | max. 50 ppm |
Total N | max. 50 ppm |
Selenate and sulfate (as SO₄) | max. 50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...