Carboxymethyl cellulose sodium salt, low viscosity, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

217277-250GM 217277-1KG
80502-040EA 121.36 USD
80502-040 80502-038
Carboxymethyl cellulose sodium salt, low viscosity, Calbiochem®
Carboxymethyl cellulose sodium salt

Substrate for cellulase.

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 14,01829

Order Now

Specification Test Results

Viscosity (2%, H₂O, 25 °C) 25 - 75 mPas

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR