2,2,6,6-Tetramethylpiperidine ≥99%, liquid
Supplier: CHEM IMPEX INTL INC MS
|
|
24504-25G
24504-1KG
24504-250G
24504-100G
24504-100G
24504-250G
24504-25G
102601-562EA
27.08
USD
102601-562
102601-564
102613-650
102625-146
CH24504-100G
CH24504-250G
CH24504-25G
2,2,6,6-Tetramethylpiperidine ≥99%, liquid
2,2,6,6-Tetramethylpiperidine
2,2,6,6-Tetramethylpiperidine can be used as a hindered base (hindered amine) because it can dissolve in organic solvents unlike inorganic bases such as potassium hydroxide.
Synonym: Norpempidine
Formula:
C₉H₁₉N MW: 141.26 g/mol Boiling Pt: 152 Density: 0.837 g/mL (25 °C) Flash Pt: 24 Storage Temperature: Freezer |
MDL Number:
MFCD00005985 CAS Number: 768-66-1 UN: 1992 ADR: 3 III |
Specification Test Results
Refractive Index (n20/D) | 1.445 |
Purity (GC) | ≥99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...