Paromomycin sulfate ≥675 mcg/mg USP

Supplier: CHEM IMPEX INTL INC MS

Synonyms: (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)

01507-5G 01507-1G
102616-218EA 153.98 USD
102616-218 102643-190
Paromomycin sulfate ≥675 mcg/mg USP
Paromomycin sulfate
Formula: C₂₃H₄₅N₅O₁₄·H₂SO₄
MW: 615.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079278
CAS Number: 1263-89-4

Order Now

Specification Test Results

Optical Rotation +50 – +55° (C=1 in H2O)
Loss on Drying ≤5.0%
Residue on Ignition ≤2.0%
Potency (Anhydrous Basis) 675 mcg/mg
pH 5.0 – 7.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR