DuPont™ Tychem® 10000 Coveralls with Respirator Hood and Attached Socks
Supplier: DuPont
TK128TLY4X000200 TK128TLY5X000200 TK128TLY6X000200 TK128TLYLG000200 TK128TLY2X000200 TK128TLY3X000200 TK128TLYMD000200 TK128TLYSM000200 TK128TLYXL000200
76187-666CS 436.32 USD
76187-666 76187-668 76187-670 76187-672 76187-878 76187-880 76187-882 76187-884 76187-886
DuPont™ Tychem® 10000 Coveralls with Respirator Hood and Attached Socks
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Respirator fit hood
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Double storm flap with adhesive closure
  • Taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached respirator-fit hood with elastic around face opening is designed to cover neck and chin and fit around respirator face mask. Strong zipper allows for tough, flexible closure. Longer zipper extends to chin for complete coverage of neck area. Storm flap covers zipper, which can be sealed by the wearer with two adhesive strips to prevent intrusion at zipper. Elastic opening at wrist allows for a tighter fit. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certificaciones: Thickness meets ASTM D1777. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.