Prostaglandin I Metabolite ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
501100-480 501100-96
75816-854EA 1501.17 USD
75816-854 75816-856
Prostaglandin I Metabolite ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of PGIM in urine and other sample matrices.
  • Measure PGIMs in urine
  • Measure PGIM down to 120 pg/mL
  • Convenient overnight format
Prostaglandin I (Prostaglandin I2; PGI2) is a potent vasodilator and inhibitor of platelet aggregation that is formed from arachidonic acid primarily in the vascular endothelium and renal cortex by sequential activities of cyclooxygenase (COX) and PGI2 synthase. It is non-enzymatically hydrated to 6-keto PGF (t½ = 2-3 minutes), and then quickly converted to numerous downstream metabolites, collectively known as prostaglandin I metabolites or PGIM, (t½= 30 minutes). Although 6-keto PGF is commonly measured in plasma and urine as an estimate of PGI2 synthesis, it should be noted that there may be more than one source of PGI2 in these samples. For instance, urinary concentrations of 6-keto PGF are confounded by the fact that only a portion originates from plasma and the remainder is produced by the kidney. These problems are circumvented by measuring downstream PGIM rather than 6-keto PGF as an indicator of systemic PGI2 production. Cayman's PGIM ELISA Kit is a competitive assay that can be used for quantification of PGIM in urine and other sample matrices. The assay has a range from 39 – 5000 pg/mL and a sensitivity (80% B/B0) of approximately 120 pg/mL.