DuPont™ Tychem® 6000 Coveralls with Respirator Hood
Supplier: DuPont
TF145TGY2X000600 TF145TGY2X0006TV TF145TGY3X000600 TF145TGY3X0006TV TF145TGY4X000600 TF145TGY4X0006TV TF145TGY5X000600 TF145TGY5X0006TV TF145TGY6X000600 TF145TGY7X000600 TF145TGYLG000600 TF145TGYLG0006TV TF145TGYMD000600 TF145TGYMD0006TV TF145TGYSM000600 TF145TGYSM0006TV TF145TGYXL000600 TF145TGYXL0006TV
76187-618CS 432.95 USD
76187-618 76187-620 76187-622 76187-624 76187-626 76187-628 76187-630 76187-632 76187-634 76187-636 76187-638 76187-640 76187-642 76187-644 76187-646 76187-648 76187-650 76187-652
DuPont™ Tychem® 6000 Coveralls with Respirator Hood
Disposable Garments
Tychem® 6000 garments are made of a proprietary barrier film laminated to a Tyvek® substrate.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap
  • Taped seams
The fabric provides at least 30 minutes of protection against over 181 challenge chemicals including chemical warfare agents and toxic industrial chemicals. Tychem® 6000 garments are strong, durable and lightweight, and they are available in both high-visibility orange and low-visibility gray fabrics, making them a preferred choice for law enforcement, emergency medical services (EMS)More Product Information