DuPont™ ProShield® 50 Coveralls with Standard Hood and Attached Skid Resistant Boots
Supplier: DuPont
NB122SWHMD002500 NB122SWHSM002500 NB122SWHXL002500 NB122SWH2X002500 NB122SWH3X002500 NB122SWH4X002500 NB122SWH5X002500 NB122SWH6X002500 NB122SWHLG002500
76187-522CS 142.05 USD
76187-522 76187-524 76187-526 76187-814 76187-816 76187-818 76187-820 76187-822 76187-824
DuPont™ ProShield® 50 Coveralls with Standard Hood and Attached Skid Resistant Boots
Disposable Garments
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Standard fit hood
  • Elastic wrists
  • Attached skid-resistant boots
  • Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include: janitorial, sanitation, and general industrial maintenance.
Certificaciones: Thickness meets ASTM D751.