BriskHeat® Silicone Rubber Heating Tapes with Adjustable Time Percentage Control, Cole-Parmer
Supplier: Antyila Scientific
03106-32 03106-33 03106-34 03106-35 03106-36
76181-516EA 161.99 USD
76181-516 76181-518 76181-520 76181-522 76181-524
BriskHeat® Silicone Rubber Heating Tapes with Adjustable Time Percentage Control, Cole-Parmer
Tapes
Regulates the length of time the heater is on and off.
  • Heat to 450 °F (232 °C)
  • Flexible silicone rubber is moisture resistant
Solve a wide range of heating applications without risk of damage due to chemical and moisture spillover. Tapes are available with either built-in time percentage control. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in highstrength, high-temperature-resistant silicone rubber.
These tapes are ideal for tight radius heating applications and are chemical, abrasion and moisture resistant.
Precaución: Heating tapes should not be overlapped.