Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]

Supplier: Biotium

BNUB1000-100 BNUB1000-500
75886-090EA 240.92 USD
75886-090 75886-092
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]
Antibodies
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone BRD469 BRD494 BRD.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR