Anti-BrdU Mouse Monoclonal Antibody [clone: 85-2C8]

Supplier: Biotium

BNUB0886-100 BNUB0886-500
75882-456EA 240.92 USD
75882-456 75882-458
Anti-BrdU Mouse Monoclonal Antibody [clone: 85-2C8]
Antibodies
This antibody recognizes Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone 85-2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR