Anti-ACTA2 Mouse Monoclonal Antibody (Biotin) [clone: ACTA2/791 1A4]

Supplier: Biotium

BNCB1203-100 BNCB1203-500
75974-184EA 205.77 USD
75974-184 75974-186
Anti-ACTA2 Mouse Monoclonal Antibody (Biotin) [clone: ACTA2/791 1A4]
Antibodies
Actin is a major component of the cytoskeleton and is present in most cell types. It is highly specific to actin from smooth muscles. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: Clone ACTA2/791 1A4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR