Anti-ACTA2 Mouse Monoclonal Antibody (CF640R) [clone: ACTA2/791 1A4]
Supplier: Biotium
BNC401203-100 BNC401203-500
75973-120EA 205.77 USD
75973-120 75973-122
Anti-ACTA2 Mouse Monoclonal Antibody (CF640R) [clone: ACTA2/791 1A4]
Antibodies
Actin is a major component of the cytoskeleton and is present in most cell types. It is highly specific to actin from smooth muscles. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actinMore Product Information