Anti-HSP90B1 Rat Monoclonal Antibody (CF568) [clone: Clone HSP90B1/1192]

Supplier: Biotium

BNC681192-100 BNC681192-500
75949-350EA 205.77 USD
75949-350 75949-352
Anti-HSP90B1 Rat Monoclonal Antibody (CF568) [clone: Clone HSP90B1/1192]
Antibodies
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: Clone HSP90B1/1192
Conjugation: CF568
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR