Anti-CHGA Mouse Monoclonal Antibody (Biotin) [clone: CGA414 CGA/777 CGA/798]

Supplier: Biotium

BNCB1060-100 BNCB1060-500
75910-080EA 205.77 USD
75910-080 75910-082
Anti-CHGA Mouse Monoclonal Antibody (Biotin) [clone: CGA414 CGA/777 CGA/798]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: CGA414 CGA/777 CGA/798
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Monkey, Mouse, Pig, Ra
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR