Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: BCL2/782 BCL2/796]

Supplier: Biotium

BNC401023-500 BNC401023-100
75882-264EA 569.73 USD
75882-264 75884-894
Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: BCL2/782 BCL2/796]
Antibodies
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: Clone BCL2/782 BCL2/796
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR