Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: HepPar1]

Supplier: Biotium

BNUB0966-100 BNUB0966-500
75951-576EA 240.92 USD
75951-576 75951-578
Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: HepPar1]
Antibodies
Hepatocyte Paraffin 1 or HepPar1 localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. The Hepatocyte Specific Antigen may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of the Hepatocyte Specific Antigen correlates with smaller tumor size and longer patient survival. Occasionally, Hepatocyte Specific Antigen is also found in gastric carcinomas as well as in a few other non-hepatic tumors.

Type: Primary
Antigen: Hepatocyte Specific
Clonality: Monoclonal
Clone: Clone HepPar1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR