Anti-CD47 Mouse Monoclonal Antibody (Biotin) [clone: IAP/964]

Supplier: Biotium

BNCB0964-100 BNCB0964-500
75913-790EA 205.77 USD
75913-790 75913-792
Anti-CD47 Mouse Monoclonal Antibody (Biotin) [clone: IAP/964]
Antibodies
This antibody reacts with Ig domain of CD47 protein. CD47, originally named integrin-associated protein (IAP), is a 50 kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: Clone IAP/964
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR