Anti-BSG Mouse Monoclonal Antibody (Biotin) [clone: BSG/963]

Supplier: Biotium

BNCB0963-100 BNCB0963-500
75887-730EA 205.77 USD
75887-730 75887-732
Anti-BSG Mouse Monoclonal Antibody (Biotin) [clone: BSG/963]
Antibodies
This MAb recognizes extracellular epitope of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: Clone BSG/963
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR