Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: B2M/961]

Supplier: Biotium

BNC680961-100 BNC680961-500
75884-266EA 205.77 USD
75884-266 75884-268
Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: B2M/961]
Antibodies
This antibody recognizes a protein of 12 kDa, identified as β2-microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of analpha heavy chain that contains three subdomains (α1,alpha2,alpha3) and a non-covalent associating light chain, known as beta-2-Microglobulin. beta-2-Microglobulin associates with thealpha3 subdomain of thealpha heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. Thealpha1 andalpha2 domains of thealpha heavy chain form the peptide antigen-binding cleft. Mutations in the beta-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: Clone B2M/961
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Non-human primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR