Anti-HSP90B1 Rat Monoclonal Antibody (CF640R) [clone: Clone 9G10.F8.2]

Supplier: Biotium

BNC400940-100 BNC400940-500
75949-586EA 205.77 USD
75949-586 75949-588
Anti-HSP90B1 Rat Monoclonal Antibody (CF640R) [clone: Clone 9G10.F8.2]
Antibodies
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: Clone 9G10.F8.2
Conjugation: CF640R
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Chicken, Cow, Dog, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR