Anti-GFAP Mouse Monoclonal Antibody (Biotin) [clone: GA-5 ASTRO/789]

Supplier: Biotium

BNCB0898-100 BNCB0898-500
75927-564EA 205.77 USD
75927-564 75927-566
Anti-GFAP Mouse Monoclonal Antibody (Biotin) [clone: GA-5 ASTRO/789]
Antibodies
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5 ASTRO/789
Conjugation: Biotin
Epitope:
Host: Mouse
Reactivity: Chicken, Cow, Human, Mouse, Pig, Rabbit, Rat
Isotype: IgGs
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR