Anti-TYRP1 Mouse Monoclonal Antibody (CF568) [clone: TA99 TYRP1/807]

Supplier: Biotium

BNC680808-100 BNC680808-500
75977-972EA 205.77 USD
75977-972 75977-974
Anti-TYRP1 Mouse Monoclonal Antibody (CF568) [clone: TA99 TYRP1/807]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: Clone TA99 TYRP1/807
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR