Anti-KRT10 Mouse Monoclonal Antibody (CF405S) [clone: DE-K13]

Supplier: Biotium

BNC040696-100 BNC040696-500
75918-314EA 205.77 USD
75918-314 75918-316
Anti-KRT10 Mouse Monoclonal Antibody (CF405S) [clone: DE-K13]
Antibodies
This antibody recognizes cytokeratin 10 (56.5 kDa) and cytokeratin 13 (53 kDa) in Western blotting. It recognizes only cytokeratin 13 in formalin-fixed, paraffin-embedded tissue sections. It does not react with cytokeratin 10 positive, cytokeratin 13 negative epithelia such as epidermis. However, on frozen sections this MAb serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and non-cornifying stratified epithelia and more differentiated cells of squamous carcinomas.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: Clone DE-K13
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR