Anti-CSF2 Rat Monoclonal Antibody (CF594) [clone: Clone BVD2-21C11]

Supplier: Biotium

BNC940662-100 BNC940662-500
75948-814EA 205.77 USD
75948-814 75948-816
Anti-CSF2 Rat Monoclonal Antibody (CF594) [clone: Clone BVD2-21C11]
Antibodies
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: Clone BVD2-21C11
Conjugation: CF594
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR