Anti-EGFR Mouse Monoclonal Antibody (CF568) [clone: H9B4]

Supplier: Biotium

BNC680642-100 BNC680642-500
75925-876EA 205.77 USD
75925-876 75925-878
Anti-EGFR Mouse Monoclonal Antibody (CF568) [clone: H9B4]
Antibodies
This MAb reacts with a cytoplasmic domain of EGFR. EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin & epiregulin.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: Clone H9B4
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR