Anti-EGFR Mouse Monoclonal Antibody (CF640R) [clone: H9B4]

Supplier: Biotium

BNC400642-100 BNC400642-500
75925-668EA 205.77 USD
75925-668 75925-670
Anti-EGFR Mouse Monoclonal Antibody (CF640R) [clone: H9B4]
Antibodies
This MAb reacts with a cytoplasmic domain of EGFR. EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin & epiregulin.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: Clone H9B4
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR