Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER506]

Supplier: Biotium

BNCB0506-100 BNCB0506-500
75926-184EA 205.77 USD
75926-184 75926-186
Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER506]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ER506
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR