Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER505]

Supplier: Biotium

BNCB0505-100 BNCB0505-500
75924-740EA 205.77 USD
75924-740 75924-742
Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER505]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ER505
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR